Learn More
Teknova 150mM PBS, pH 6.0. 5L, Sterile 2 pack.
SDP
Supplier:  Teknova P38502PK
Encompass
Phosphate-buffered saline (PBS) is a salt solution that is commonly used in physiological and biochemical experiments. It can be used to maintain pH neutrality or to buffer against changes in pH in order to provide optimal conditions for cellular growth and function. The buffering range is between pH 6.8 and 7.4.
Catalog No. 50-274-5894