Learn More
Research Products International Corp LB Brth Lnnx Pwd Anml Free
SDP
Supplier:  Research Products International Corp L243601000.0
Encompass_Preferred
For propagation and maintenance of E. Coli. Low Salt Formula. Molecular Biology Grade. Dissolve 20 grams in 1 liter distilled water. Then adjust pH to 7.2. Heat and frequently agitate to completely dissolve. Sterilize by autoclaving for 15 minutes.
Catalog No. 50-297-9108