Learn More
Kd Medical Inc Lb + Gentamycin 7Ug/ml + Kanamycin 50Ug/ml + Tetracycline 10Ug/ml + Xgal 60Ug/ml + Iptg .5mm, 100mm, 10/Sleeve
SDP
Supplier:  Kd Medical Inc BPL2550
Encompass_Preferred
This item has a minimum order quantity of 4 per supplier requirements. Used for cultivation of E.Coli that contains plasmids conferring Gentamycin (7ug/ml), Kan (50ug/ml), and Tetracyline (10ug/ml) resistance. X-Gal (60ug/ml)used for blue/white screening of colonies. IPTG 5mM.Ten 100mm plates per sleeve.
Catalog No. 50-103-1257