Learn More
FUJIFILM BIOSCIENCES INC LBIS Mouse/Rat High Molecular
Supplier:  FUJIFILM BIOSCIENCES INC 29790701
Encompass_Preferred
Adiponectin is a cytokine secreted by adipocytes and plays a role in regulating lipid metabolism and insulin sensitivity and is known for its antidiabetic anti-atherosclerotic and anti-inflammatory effects In the bloodstream adiponectin exists in multimeric forms trimers low molecular weight LMW hexamers medium molecular weight MMW and higher-order complexes of 12 to 18 monomers high molecular weight HMW Circulating HMW adiponectin levels rather than total adiponectin are considered a more accurate indicator of physiological parameters such as BMI sex weight loss glucose tolerance hepatic insulin sensitivity and the presence of metabolic syndrome or type 2 diabetes
Catalog No. 50-004-50417