Learn More
Aviva Systems Biology Corp LBX2 Antibody - middle region (ARP47507_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47507_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LBX2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LBX2. Alternate names: LP3727ApplicationWB. Species reactivity: Human, Pig
Catalog No. 50-014-59571