Learn More
Medchemexpress LLC LC-04-045 NEK7 molecular glue degrader | 99.6% | 566.05 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY172162100MG
Encompass_Preferred
LC-04-045 is a selective NIMA-related kinase 7 (NEK7) molecular glue degrader. It induces NEK7 degradation via the CRBN-based ubiquitin-proteasome system, dependent on the glycine 57-containing degron motif. This product inhibits the secretion of downstream cytokines IL-1β and IL-18 and can be used for research on lymphoblastic leukemia.
  • Selective NEK7 molecular glue degrader
  • Induces NEK7 degradation via CRBN-based ubiquitin-proteasome system
  • Inhibits secretion of IL-1β and IL-18
  • Suitable for lymphoblastic leukemia research
  • Purity of 99.56%
Catalog No. 50-003-86369