Learn More
Takara Bio Living Colors Full-Length ZsGreen Polyclonal Antibody, 100 uL
SDP
Supplier:  Takara Bio 632474
Encompass
The Living Colors Full-Length ZsGreen Polyclonal Antibody was raised in a rabbit against recombinant full-length Zoanthus sp. green fluorescent protein (ZsGreen). This antibody recognizes native and denatured forms of ZsGreen as well as fusion proteins containing ZsGreen in bacterial and mammalian cell lysates.
Catalog No. NC1250698