Learn More
Cayman Chemical LC3-mHt-IN-n1 10mg
SDP
Supplier:  Cayman Chemical 3430610 MG
Encompass_Preferred
An LC3B-mutant huntingtin linker; decreases HdhQ7/Q140 levels in primary mouse cortical neurons at 300 nM; increases survival of Drosophila expressing human HdhQ128
Catalog No. 50-294-7719