Learn More
ABclonal Technology LCAT Rabbit pAb
SDP
Supplier:  ABclonal Technology A1174120UL
Encompass_Preferred
This gene encodes the extracellular cholesterol esterifying enzyme, lecithin-cholesterol acyltransferase. The esterification of cholesterol is required for cholesterol transport. Mutations in this gene have been found to cause fish-eye disease as well as LCAT deficiency.
Catalog No. 50-001-16226