Learn More
Selleck Chemical LLC LCL161
SDP
Supplier:  Selleck Chemical LLC S700925MG
Encompass_Preferred
LCL-161 a small molecule second mitochondrial activator of caspase (SMAC) mimetic potently binds to and inhibits multiple IAPs (i e XIAP c-IAP)
Catalog No. 50-000-02237