Learn More
Selleck Chemical LLC LCL161 5mg 1005342-46-0
SDP
Supplier:  Selleck Chemical LLC S70095MG
Encompass_Preferred
LCL-161, a small molecule second mitochondrial activator of caspase (SMAC) mimetic, potently binds to and inhibits multiple IAPs (i.e. XIAP, c-IAP). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4319