Learn More
Medchemexpress LLC LCMV gp33-41 TFA | 98.5% | 1158.25 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYP1569A50MG
Encompass_Preferred
LCMV gp33-41 (TFA), a carboxyl-extended 9-aa-long peptide, is a lymphocytic choriomeningitis virus sequence restricted by MHC class I H-2Db molecules and presented to cytotoxic T lymphocytes. It is located at nucleotides 175-201 of the S segment of the LCMV genome, can be presented by the mouse MHC I molecule H-2Db and recognized by CD8+ T cells. It accumulates multiple non-synonymous mutations during chronic infection, which may help the virus evade clearance by the immune system.
  • Carboxyl-extended 9-aa-long peptide
  • Lymphocytic choriomeningitis virus sequence
  • Restricted by MHC class I H-2Db molecules
  • Presented to cytotoxic T lymphocytes
  • Recognized by CD8+ T cells
  • Accumulates non-synonymous mutations during chronic infection
  • Supports evasion of virus clearance by the immune system
  • For research use only
Catalog No. 50-004-24302