Learn More
Apexbio Technology LLC LDC000067, 50mg. Cas: 1073485-20-7 MFCD: MFCD28137788. CDK9 inhibitor, novel and highly specific
SDP
Supplier:  Apexbio Technology LLC B475450
Encompass_Preferred
LDC000067 (LDC067) is a novel specific inhibitor of CDK9 with IC50 value of 44 10 nM [1].Cyclin-dependent kinase 9 (CDK9) is a cyclin-dependent kinase. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5359