Learn More
Selleck Chemical LLC LDC4297 (LDC044297) 5mg 1453834-21-3
SDP
Supplier:  Selleck Chemical LLC S79925MG
Encompass_Preferred
LDC4297 is a novel CDK7 inhibitor (IC50=0.13¿0.06 nM for CDK7 versus IC50s between 10 nM and 10,000 nM for all other analyzed CDKs). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5516