Learn More
TARGETMOL CHEMICALS INC LDCA 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T604005MG
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. LDCA is a double-hit metabolic regulator that exhibits potent anti-proliferative activity. lDCA competitively inhibits LDH-A enzyme activity alters mitochondrial hyperpolarization and simultaneously subverts apoptosis. lDCA has a pronounced effect in cancer cells and does not show toxicity. purity: 98%
Catalog No. 50-257-9621