Learn More
Apexbio Technology LLC LDH Cytotoxicity Assay Kit (Fluorometric) 200T
SDP
Supplier:  Apexbio Technology LLC K2267200
Encompass_Preferred
LDH Cytotoxicity Assay Kit Fluorometric is a biochemical assay designed to assess cytotoxicity by quantifying lactate dehydrogenase (LDH) activity released from damaged or compromised cells It enables evaluation of cell injury or death through measurement of LDH-catalyzed conversion of lactate to pyruvate resulting in NAD reduction to NADH The resulting NADH interacts with assay reagents to produce a fluorescent signal allowing quantitative detection of cell damage via fluorescence intensity This fluorometric assay serves as a non-radioactive alternative to chromium-51 release assays The kit is suitable for research applications involving cytotoxicity assessment
Catalog No. 50-000-97291