Learn More
ABclonal Technology LDHA Rabbit pAb
SDP
Supplier:  ABclonal Technology A11461000UL
Encompass_Preferred
The protein encoded by this gene catalyzes the conversion of L-lactate and NAD to pyruvate and NADH in the final step of anaerobic glycolysis. The protein is found predominantly in muscle tissue and belongs to the lactate dehydrogenase family. Mutations in this gene have been linked to exertional myoglobinuria. Multiple transcript variants encoding different isoforms have been found for this gene. The human genome contains several non-transcribed pseudogenes of this gene.
Catalog No. 50-001-15275