Learn More
Cayman Chemical LDN-192960hydrochlorIde 5mg
SDP
Supplier:  Cayman Chemical 190895 MG
Encompass_Preferred
An inhibitor of haspin and DYRK2 (IC50s = 10 and 48 nM, respectively); selective for haspin and DYRK2 over TRKB, CLK1, DYRK1A, DYRK3, ROS, HIPK1, HIPK2, PIM-1, and PIM-2 kinases (IC50s = 720-91,000 nM)
Catalog No. 50-298-3053