Learn More
Selleck Chemical LLC LDN-193189 2mg 1062368-24-4 DM3189
SDP
Supplier:  Selleck Chemical LLC S26182MG
Encompass_Preferred
LDN-193189 (DM3189) is a selective BMP signaling inhibitor, inhibits the transcriptional activity of the BMP type I receptors ALK2 and ALK3 with IC50 of 5 nM and 30 nM in C2C12 cells, respectively, exhibits 200-fold selectivity for BMP versus TGF-?. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0225637