Learn More
Apexbio Technology LLC LDN193189 Hydrochloride 1062368-62-0 10mg
SDP
Supplier:  Apexbio Technology LLC A354510
Encompass_Preferred
LDN193189 Hydrochloride is a selective pharmacological inhibitor targeting BMP type I receptor kinases specifically activin receptor-like kinase 2 (ALK2) and ALK3 The compound acts by blocking receptor-mediated activation of Smad-dependent signaling pathways (Smad1/5/8 phosphorylation) as well as mitigating associated non-Smad signaling including phosphorylation of Akt and p38 Commonly utilized in biomedical research involving cellular differentiation epithelial barrier integrity and heterotopic ossification LDN193189 has shown the capacity to modulate BMP-induced phenotypic changes in various cell lines such as C2C12 myoblasts and bronchial epithelial cells and to influence related in vivo processes
Catalog No. 50-000-63245