Learn More
NABERTHERM INC LE1/11/R7,TMAX 1100C 120V
Supplier:  NABERTHERM INC LE013K17N1120
Encompass_Preferred
This economy muffle furnace is designed for many lab applications. Features: Tmax 1100 °C, working temp. 1050 °C, heating from two sides from heating elements in quartz glass tubes, dual shell furnace housing of rust-free stainless steel, Insulation made of non-classified fiber material
Catalog No. NC2068355