Learn More
TARGETMOL CHEMICALS INC Leachianone A 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC TN18565MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg and bulk. Please contact Fisher for quotes.Leachianone A (Isokurarinone) is a potential antitoxic agent, it shows inhibitory effects on cadmium- Induced cytotoxicity. Purity 99.79%
Catalog No. 50-288-0279