Learn More
CTL Scientific Supply Corp LEAD ACETATE PAPER,PK/4 BOOKS
SDP
Supplier:  CTL Scientific Supply Corp 907464
Encompass_Preferred
LEAD ACETATE PAPER - Pack of 4 booklets of 100 strips. Limit of sensitivity: 1 drop of a solution of 5 mg/liter sulfide gives a brown ring, which is just visible.
Catalog No. 50-117-3305