Learn More
Research Products International Corp Lead Apron, Versa-Adjustable, 24 Inches Wide, 36 Inches Long, Beige
SDP
Supplier:  Research Products International Corp 103538B
Encompass
Features two hook & loop fastening flaps at the waist, which aid in relieving weight off the shoulders and includes adjustable padded shoulders. Its comfortable fit is attained by varying the position of the flaps. Shoulder pads included.
Catalog No. 50-213-075