Learn More
Vee Gee Lead C Ring 125-500mL flasks
Supplier:  Vee Gee HS8881A
Encompass_Preferred
The Lead Ring is perfect for stabilizing vessels and labware or for water bath immersions.* Vinyl-coated lead ring will not mark or scratch benchtops and labware* C-Shape Style allows some manual size variation for a customized fit
Catalog No. 50-288-6377