Learn More
Electron Microscopy Sciences Lead Citrate, Ready to use 30 ML
SDP
Supplier:  Electron Microscopy Sciences 22410
Encompass_Preferred
Use lead citrate as a counterstain with UranyLess to enhance the contrast in negative staining applications for TEM. The special pump delivers the product without letting in air, thus preserving the solution and preventing CO2 dissolution. It is convenient to use because it performs in any position, even upside down.
Catalog No. NC1588038