Learn More
Strem, An Ascensus Company CAS# 7758-95-4. 100g. Lead(II) chloride, 99%. MFCD00011157
SDP
Supplier:  Strem, An Ascensus Company 938206100G
Encompass
CAS# 7758-95-4. 100g. Lead(II) chloride, 99%. MFCD00011157. Molecular Weight: 278.10. Molecular Formula: PbCl2. Color/form: white pwdr. Strem# 93-8206. http://www.strem.com/catalog/v/93-8206/
Catalog No. 50-901-16476