Learn More
Strem, An Ascensus Company CAS# 7783-46-2. 500g. Lead(II) fluoride, 99+%. MFCD00011162
SDP
Supplier:  Strem, An Ascensus Company 938214500G
Encompass
CAS# 7783-46-2. 500g. Lead(II) fluoride, 99+%. MFCD00011162. Molecular Weight: 245.19. Molecular Formula: PbF2. Color/form: white pwdr. Strem# 93-8214. http://www.strem.com/catalog/v/93-8214/
Catalog No. 50-901-16481