Learn More
Strem, An Ascensus Company CAS# 10101-63-0. 50g. Lead(II) iodide (99.999+%-Pb) PURATREM. MFCD00011163
SDP
Supplier:  Strem, An Ascensus Company 82075050G
Encompass
CAS# 10101-63-0. 50g. Lead(II) iodide (99.999+%-Pb) PURATREM. MFCD00011163. Molecular Weight: 461.00. Molecular Formula: PbI2. Color/form: yellow pwdr. Strem# 82-0750. http://www.strem.com/catalog/v/82-0750/
Catalog No. 50-901-13519