Learn More
BioSpec Products LEAF DISC PUNCHER, 1/4 INCH DI
SDP
Supplier:  BioSpec Products 709
Encompass_Preferred
The leaf tissue punch produces a single 1/4 inch dia. disc. The punch holds a standard snap-top or screw-top microvial in a position to receive the sample without further manipulation.
Catalog No. NC0769832