Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lecanoric acid | 480-56-8 | 5 MG
SDP

Supplier:  Medchemexpress LLC HYN33945MG

Encompass_Preferred

Lecanoric acid is a histidine-decarboxylase inhibitor isolated from fungus. Its inhibition is competitive with histidine and noncompetitive with pyridoxal phosphate. Lecanoric acid does not inhibit aromatic amino acid decarboxylase.

  • Molecular weight: 318.28
  • Formula: C16H14O7
  • Appearance: Solid
  • Color: White to off-white
  • Structure classification: Phenols, polyphenols
  • Initial source: Microorganisms (Piricularia strain FI-178)
  • Purity: 98.0%
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: 4°C, protect from light. In solvent: -80°C for 6 months; -20°C for 1 month (protect from light)

Catalog No. 50-004-19076


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.