Learn More
TARGETMOL CHEMICALS INC LECIMIBIDE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T256515MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Lecimibide (DuP 128) is a potent and selective inhibitor of acyl coenzyme A cholesterol acyltransferase (ACAT) which can be used to study diseases due to high lipids. purity: 99%
Catalog No. 50-258-0744