Learn More
Medchemexpress LLC Lecirelin | 61012-19-9 | 99.6% | 1209.40 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYP005150MG
Encompass_Preferred
Lecirelin is a synthetic gonadotropin-releasing hormone (GnRH) analogue that acts as a GnRH agonist. It is widely used for the research of bovine ovarian follicular cysts. In studies, Lecirelin (2.07 μM) significantly increases the tension and frequency of contractions in preovulatory follicles (PF), small follicular cysts (FC-S), and large follicular cysts (FC-L). It also causes strips from preovulatory follicles to contract significantly more than strips from cysts.
  • Synthetic gonadotropin-releasing hormone (GnRH) analogue
  • Acts as a GnRH agonist
  • Used in research of bovine ovarian follicular cysts
  • Increases tension and frequency of contractions in preovulatory follicles
  • Increases tension and frequency of contractions in small follicular cysts
  • Increases tension and frequency of contractions in large follicular cysts
Catalog No. 50-004-26520