Learn More
Medchemexpress LLC Lecithin | 8002-43-5 | 98.0% | 5 G
SDP
Supplier:  Medchemexpress LLC HYB22355G
Encompass_Preferred
Lecithin is considered a safe, traditional source of phospholipid. Phospholipids are known to modify the fatty acid composition and microstructure of membranes in animal cells.
  • Regarded as a safe, conventional phospholipid source.
  • Phospholipids alter the fatty acid composition and microstructure of membranes in animal cells.
  • Enhances bile resistance in Lactobacillus plantarum strains by soy lecithin.
  • Increases the survival rate of harvested cells in bile challenge.
  • Enhances cell surface hydrophobicity and membrane integrity.
Catalog No. 50-004-10561