Learn More
Dazor Mfg Corporation LED Circline Desk Base 3 Diopter Magnifier Providing 1.75X magnification, 5" Round Lens . 33” Arm Reach. Color: White
Supplier:  Dazor Mfg Corporation L1470WH
Encompass_Preferred
Dazor's LED Circline Magnifier is the ultimate tool for providing the perfect amount of magnification with abundant, shadow-free light. The ideal all-purpose magnifier.FeaturesProvides abundant, even and shadow-free light over a broad surfaceInnovative Glow Ring diffuses and softens the light emitted by the 144 LEDs and simulates the light output of a traditional fluorescent bulbTwice as bright as a 22W fluorescent bulb yet fully dimmable to 20% intensitySeparate ON/OFF and dimming controlsFeatures Dazor's patented 33" floating adjustable arm for smooth and easy positioning of the lamp to any positionSuperior energy-efficiencySturdy weighted desk baseDurable powder-coat finish5 year mechanical / 2 year electrical warrantyMade in the USA
Catalog No. NC1428419