Learn More
Leica Biosystems LED Lamp Housing for Leica DM6 Microscope, Long Cable
Supplier:  Leica Biosystems 11504242
Encompass_Preferred
LED lamp housing designed for use with the DM6 upright microscope series. The module provides LED-based illumination and includes an extended-length cable for flexible installation and routing.
  • Provides LED-based illumination for microscope observation and imaging.
  • Includes an extended-length cable for flexible installation and routing.
  • Designed as a replacement lamp housing for DM6 upright microscopes.
  • Solid-state LED light source for stable output and long operational lifetime.
  • Intended for use in laboratory, materials, and metallographic microscopy applications.
Catalog No. NC3365988