Learn More
Microscope World SCHOTT ColdVision CV-LS LED Light Source
SDP
Supplier:  Microscope World CVLS
Encompass_Preferred
This SCHOTT ColdVision CV-LS LED Light Source, cool white 6000K light box features internal light feedback stabilization, fast triggering strobe.Color Temperature- 6000K cool white.Luminous Flux- Up to 13502 lm light flux.Max Active Light Guide Ø- 13mmVoltage- 100-240VLight Box- Continuous dimming, fan cooling, ESD Immunity.Analog and Digital Remote Control- USB, RS-232, Dual Ethernet.Light Box Dimensions / Weight- 129mm x 201mm x 59.7mm (5.08inch x 7.93inch x 2.35inch), 0.816kg (1.8 lb)Certifications- CEWarranty- 2 years
Catalog No. NC2965661