Learn More
Microscope World SCHOTT ColdVision MC-LS with 38-53mm Ring Light
SDP
Supplier:  Microscope World MCLSR3853
Encompass_Preferred
This SCHOTT ColdVision MC-LS LED Light Source with a ring light is designed for use in stereo microscopes 38-53mm in diameter. Ring light specifications-Objective diameter range- 38 to 53mmInner diameter- 53.1mm (2.09inch)Outer diameter- 58.7mm (2.31inch)Working distance range- 13 to 38 mm (0.5inch to 1.5inch)Length of connection to box- 1016mm (40inch)ColdVision MC-LS Specifications-Color Temperature- 5400K cool white.Luminous Flux- Up to 8502 lm light flux.Max Active Light Guide Ø- 13mmVoltage- 100-240V, 50/60HzLight Box- Continuous dimming, fan cooling, temperature monitoring for protection of the LED light engine, high resistance to vibration and shock.Analog and Digital Remote Control- USB, RS-232, 9-pin connector.Light Box Dimensions / Weight- 158mm x 145mm x 98mm (6.2inch x 5.7inch x 3.9inch), 2kg (4.62 lb)Certifications- ETL Approved, RoHS compliant.Warranty- 2 years
Catalog No. NC2965655