Learn More
Microscope World SCHOTT ColdVision CV-LS with 66mm Ring Light
SDP
Supplier:  Microscope World CVLSR66
Encompass_Preferred
This SCHOTT ColdVision CV-LS LED Light Source with a ring light is designed for use in stereo microscopy and can be used with microscope objectives 66mm in diameter. The ring light fits ZEISS stereo microscope objectives. The system includes the CV-LS LED light source and a ring light.Ring light specifications-Objective diameter range- 66mmWorking distance range- 38 to 152 mm (1.5in to 6in)Length of connection to box- 838mm (33in)CV-LS Specifications-Color Temperature- 6000K cool white.Luminous Flux- Up to 13502 lm light flux.Max Active Light Guide Ø- 13mmVoltage- 100-240VLight Box- Continuous dimming, fan cooling, ESD Immunity.Analog and Digital Remote Control- USB, RS-232, Dual Ethernet.Light Box Dimensions / Weight- 129mm x 201mm x 59.7mm (5.08in x 7.93in x 2.35in), 0.816kg (1.8 lb)Certifications- CEWarranty- 2 years
Catalog No. NC2965668