Learn More
Medchemexpress LLC HY-15777 100mg , LEE011 Ribociclib CAS:1211441-98-3 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY15777100MG
Encompass_Preferred
Medchemexpress, HY-15777 100mg LEE011 Ribociclib CAS:1211441-98-3 Formula:C23H30N8O IC50: 10/39 nM (CDK4/6) Purity:98% LEE011 is a highly specific CDK4/6 inhibitor with IC 50 values of 10 nM and 39 nM, respectively, and is over 1,000-fold less potent against the cyclin B/CDK1 complex. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2666