Learn More
Selleck Chemical LLC Ribociclib (LEE011) 5mg 1211441-98-3
SDP
Supplier:  Selleck Chemical LLC S74405MG
Encompass_Preferred
Ribociclib (LEE011) is an orally available, and highly specific inhibitor of CDK4 and CDK6 with IC50 of 10 nM and 39 nM. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0871317