Learn More
Medchemexpress LLC Leelamine hydrochloride | 16496-99-4 | 98.1% | 321.93 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY110028100MG
Encompass_Preferred
Leelamine hydrochloride is a tricyclic diterpene molecule extracted from the bark of pine trees. It acts as a cannabinoid receptor type 1 (CB1) agonist and inhibits SREBP1-regulated fatty acid/lipid synthesis in prostate cancer cells, independent of androgen receptor status. It also suppresses the transcriptional activity of the androgen receptor, which is known to regulate fatty acid synthesis.
  • Tricyclic diterpene molecule
  • Extracted from the bark of pine trees
  • Cannabinoid receptor type 1 (CB1) agonist
  • Inhibits SREBP1-regulated fatty acid/lipid synthesis in prostate cancer cells
  • Suppresses transcriptional activity of androgen receptor
Catalog No. 50-003-44984