Learn More
Cayman Chemical Leelamine hydrochloride
SDP
Supplier:  Cayman Chemical 100086145 MG
Encompass_Preferred
The hydrochloride of leelamine, an inhibitor of pyruvate dehydrogenase kinase.
Catalog No. 50-288-8636