Learn More
Medchemexpress LLC HY-B0083 100mg Medchemexpress, Leflunomide CAS:75706-12-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0083100MG
Encompass_Preferred
Medchemexpress, HY-B0083 100mg Leflunomide CAS:75706-12-6 Leflunomide is a pyrimidine synthesis inhibitor, inhibiting dihydroorotate dehydrogenase, and acts as a disease-modifying antirheumatic drug. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9384