Learn More
Biolegend LEGENDplex™ Human PAI-1 Capture Bead A5, 13X
Supplier:  Biolegend 740780
Encompass_Preferred
LEGENDplex™ Human PAI-1 Capture Bead A5, 13X Reactivity: Human; Apps: Multiplex - Component; Size: 100 tests (270 uL)
Catalog No. 50-208-8368