Learn More
Biolegend LEGENDplexTM Human sTREM-1 Cap
Supplier:  Biolegend 741533
Encompass_Preferred
LEGENDplex Human sTREM-1 Capture Bead B6 13X Reactivity Human Apps Multiplex - Component Size 100 tests (270 L)
Catalog No. 50-310-7954