Learn More
Medchemexpress LLC Legumain inhibitor 1 | 2361157-34-6 | 98.3% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY12861750MG
Encompass_Preferred
Legumain inhibitor 1 is a potent and selective Legumain inhibitor with an IC50 of 3.6 nM. It is primarily used for cancer research and demonstrates high microsomal stability, with half-lives exceeding 8 hours in both mouse and human microsomes (CLint > 3 μL/min/mg).
  • Potent and selective Legumain inhibitor
  • IC50 of 3.6 nM
  • Suitable for cancer research
  • High microsomal stability with half-lives exceeding 8 hours in mouse and human microsomes
Catalog No. 50-003-42903