Learn More
Leica Biosystems Leica EM CTD Cryo tool dryer
Supplier: Leica Biosystems 16707000001
The Leica EM CTD Cryo tool dryer is a benchtop unit that rapidly de-ices and dries tools used in cryo sample preparation, combining heated airflow and a heating plate to reach working temperature quickly while protecting delicate components.
- Combines heated airflow and a heated plate for thorough drying.
- Reaches approximately 45 °C in five minutes under 20 °C ambient conditions.
- Maximum operating temperature limited to 50 °C to reduce risk of burns.
- Wide input voltage range of 100-230 V at 50-60 Hz for global compatibility.
- Compact footprint and low weight for benchtop use.