Learn More
ATCC Leishmania aethiopica (MHOM/ET/72/L100 GFP) - GFP reporter strain (PRA-417)
Supplier:  ATCC PRA417
Encompass
GFP-expressing Leishmania aethiopica reporter strain for fluorescent-based research applications including parasite detection and drug screening. Strain MHOM/ET/72/L100 GFP was transfected with green fluorescent protein and was originally isolated from a human in Ethiopia in 1972.
  • GFP-expressing reporter strain for fluorescence assays.
  • Strain designation MHOM/ET/72/L100 GFP.
  • Originally isolated from a human in Ethiopia in 1972.
  • Useful for parasite detection, imaging, and drug screening workflows.
  • Characterized and provided by a biological repository for research use.
Catalog No. NC3730039