Learn More
Medchemexpress LLC LEM-14 10mg | 1814881-70-3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11434010MG
Encompass_Preferred
LEM-14 is a small-molecule research compound that selectively inhibits NSD2 and has been used in cellular and in vivo studies to evaluate effects on cancer cell apoptosis and tumor response when combined with ionizing radiation.
  • Selective NSD2 inhibitor with reported IC50 ≈ 132 μM.
  • High purity suitable for research use (98.0%).
  • Molecular formula C25H26N4O4S, molecular weight 478.56.
  • Supplied in a 10 mg vial for laboratory studies.
  • Used in preclinical cancer and fibrosis research applications.
Catalog No. 50-002-27352